missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP30 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1415
This item is not returnable.
View return policy
Description
TIP30 Polyclonal specifically detects TIP30 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| TIP30 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| 30 kDa HIV-1 TAT-interacting protein, CC3oxidoreductase HTATIP2, EC 1.1.1, EC 1.1.1.-, FLJ26963, HIV-1 Tat interactive protein 2, 30 kDa, HIV-1 Tat interactive protein 2, 30kDa, HIV-1 TAT-interactive protein 2, SDR44U1, Tat-interacting protein (30kD), TIP30short chain dehydrogenase/reductase family 44U, member 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize all three reported isoforms. Variants (NP_006401.3; NP_001091991.1; NP_001091992.1) encode the same isoform. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_006401. | |
| HTATIP2 | |
| Peptide with sequence AETEALSKLREDFR-C corresponding to N-Terminus according to NP_001091990.1, NP_006401.3, NP_001091991.1, NP_001091992.1, NP_001091993.1. | |
| 0.1 mg | |
| Apoptosis | |
| 10553 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction