Learn More
TIPIN Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98947MFV610
This item is not returnable.
View return policy
Description
TIPIN Polyclonal antibody specifically detects TIPIN in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Rabbit | |
| RUO | |
| Purified |
| 50mM Sodium Borate | |
| 0.1 mL | |
| Primary |