missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIPIN Antibody (OTI2E6), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74528UV
This item is not returnable.
View return policy
Description
TIPIN Monoclonal specifically detects TIPIN in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TIPIN | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TIPIN | |
| Full length recombinant protein of human TIPIN (NP_060328) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2E6 | |
| Western Blot, Immunohistochemistry | |
| FLJ20516, TIMELESS interacting protein, TIMELESS-interacting protein | |
| Mouse | |
| Affinity Purified | |
| DNA Repair | |
| 54962.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction