missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIPIN Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535995
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, human TIPIN-transfected 293T cells. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Required for normal progression of S-phase. Important for cell survival after DNA damage or replication stress. May be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light.
Specifications
| TIPIN | |
| Polyclonal | |
| Unconjugated | |
| TIPIN | |
| 1110005A05Rik; 1110018P21Rik; CSM3 homolog; FLJ20516; RGD1564084; TIMELESS interacting protein; TIMELESS-interacting protein; TIMELESS-interacting-like protein; tipin; Unknown (protein for MGC:127806); zgc:91928 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54962 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9BVW5 | |
| TIPIN | |
| Recombinant protein encompassing a sequence within the center region of human TIPIN. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction