missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIPRL Antibody (OTI3D11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74529H
This item is not returnable.
View return policy
Description
TIPRL Monoclonal specifically detects TIPRL in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TIPRL | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TIPRL | |
| Full length recombinant protein of human TIPRL (NP_690866) produced in HEK293T cells. | |
| 0.1 mL | |
| 261726.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3D11 | |
| Western Blot | |
| dJ69E11.3, MGC3794, putative MAPK activating protein, Putative MAPK-activating protein PM10, TIP41, TOR signaling pathway regulator-like (S. cerevisiae), TIP41-like protein, TIP41Type 2A-interacting protein, TIPTIP41, TOR signalling pathway regulator-like (S. cerevisiae) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction