Learn More
Invitrogen™ Tiragolumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558404
 View more versions of this product
Product Code. 30283371
  • € 1204.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tiragolumab is a humanized monoclonal antibody. Tiragolumab targets TIGIT and directly or indirectly activates the anti-tumor immune response by acting on the activation pathways of APC and T cells and NK cells to achieve the purpose of eliminating tumor cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tiragolumab
Recombinant Monoclonal
Unconjugated
MTIG7192A; MTIG-7192A; MTIG-7192-A; RG-6058; RO7092284; RO-7092284
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized