missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIRAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47605
This item is not returnable.
View return policy
Description
TIRAP Polyclonal specifically detects TIRAP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TIRAP | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| adapter protein wyatt, Adaptor protein Wyatt, MAL, MyD88 adapter-like protein, MyD88-2, TIR domain-containing adapter protein, toll/interleukin-1 receptor domain-containing adapter protein, toll-interleukin 1 receptor (TIR) domain containing adaptor protein, Toll-interleukin 1 receptor (TIR) domain-containing adaptor protein, Toll-like receptor adaptor protein, wyatt | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 114609 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TIRAP | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SSRWSKDYDVCVCHSEEDLVAAQDLVSYLEGSTASLRCFLQLRDATPGGAIVSELCQALSSSHCRVLLITPGFLQDPWCKYQML | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction