missing translation for 'onlineSavingsMsg'
Learn More

TIS11D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 216.00 - € 326.00

Specifications

Antigen TIS11D
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18675662
View Documents
Novus Biologicals
NBP2-97954-100ul
100 μg
€ 326.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18607242
View Documents
Novus Biologicals
NBP2-97954-50ul
50 μg
€ 216.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TIS11D Polyclonal antibody specifically detects TIS11D in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TIS11D
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
HF.10, HF10Zinc finger protein HF.10, Zfp105, zinc finger protein 35, zinc finger protein 35 (clone HF.10)
Produced in rabbits immunized with E. coli-derived Human TIS11D fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
PBS (pH 7.0)
678
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.