missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tissue alpha-L-Fucosidase/FUCA1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21436DL594
This item is not returnable.
View return policy
Description
Tissue alpha-L-Fucosidase/FUCA1 Polyclonal antibody specifically detects Tissue alpha-L-Fucosidase/FUCA1 in Human samples. It is validated for ELISA
Specifications
| Tissue alpha-L-Fucosidase/FUCA1 | |
| Polyclonal | |
| ELISA | |
| Alpha-L-fucosidase 1, Alpha-L-fucosidase I, Alpha-L-fucoside fucohydrolase 1, EC 3.2.1, EC 3.2.1.51, FUCA, fucosidase, alpha-L- 1, tissue, tissue alpha-L-fucosidase | |
| Produced in rabbits immunized with purified, recombinant Human Tissue alpha-L-Fucosidase/FUCA1 (Gln32-Lys466; Uniprot: P04066) | |
| 0.1 mL | |
| Signal Transduction | |
| 2517 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction