Learn More
Aesculap™ Tissue Forceps
Description
- Stronger than BD537R and BD539R
- Finer than BD557R and BD559R
- Includes teeth
Specifications
Specifications
| Product Type | Tissue Forceps |
| Includes | 1 x 2 teeth |
| Tip Style | Straight |
| Length (Metric) | 145 mm |