Learn More
Aesculap™ Tissue Forceps
Product Code. 15463300
Change view
Click to view available options
Tip Style:
Straight
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Style unitSize
15463300 Straight Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15463300 Supplier Aesculap™ Supplier No. BD547R
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Aesculap™ Tissue Forceps are ideal for use in surgical applications.
  • Stronger than BD537R and BD539R
  • Finer than BD557R and BD559R
  • Includes teeth
Specifications
Product Type Tissue Forceps
Includes 1 x 2 teeth
Tip Style Straight
Length (Metric) 145 mm