missing translation for 'onlineSavingsMsg'
Learn More

Tivozanib, MedChemExpress

Product Code. 16490647
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490647

Brand: Medchem Express HY10977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tivozanib (AV-951; KRN951) is a potent and selective and orally active VEGFR tyrosine kinase inhibitor with IC50是of 0.21, 0.16, 0.24 nM for VEGFR-1, VEGFR-2, VEGFR-3, respectively. Tivozanib inhibits angiogenesis and vascular permeability in tumor tissues and shows antitumor activity. Tivozanib has the potential for the research of metastatic renal cell carcinoma (RCC) .

Tivozanib (AV-951

Specifications

Chemical Name or Material Tivozanib
CAS 475108-18-0
Color Brown
Purity Grade Notes Research
Molecular Formula C22H19ClN4O5
Quantity 50 mg
Synonym AV-951 KRN951
Solubility Information DMSO : 25 mg/mL (54.96 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C=C1Cl)OC2=CC=NC3=CC(OC)=C(C=C23)OC)NC4=NOC(C)=C4
Molecular Weight (g/mol) 454.86
Formula Weight 454.86
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.