missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TJP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94303-0.02ml
This item is not returnable.
View return policy
Description
TJP3 Polyclonal antibody specifically detects TJP3 in Human samples. It is validated for Western Blot
Specifications
| TJP3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC119546, Tight junction protein 3, tight junction protein 3 (zona occludens 3), ZO-3, ZO3tight junction protein ZO-3, zona occludens 3, Zona occludens protein 3, Zonula occludens protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 710-919 of human TJP3 (NP_001254489.1). RQWLAPASRRSTRRLYAQAQKLRKHSSHLFTATIPLNGTSDTWYQELKAIIREQQTRPIWTAEDQLDGSLEDNLDLPHHGLADSSADLSCDSRVNSDYETDGEGGAYTDGEGYTDGEGGPYTDVDDEPPAPALARSSEPVQADESQSPRDRGRISAHQGAQVDSRHPQGQWRQDSMRTYEREALKKKFMRVHDAESSDEDGYDWGPATDL | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27134 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido