missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TK1 Monoclonal Antibody (C301), Invitrogen™
GREENER_CHOICE

Product Code. p-200020128
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17970743 200 μL 200µL
17918622 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17970743 Supplier Invitrogen™ Supplier No. MA551065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thymidine Kinase (TK1) is a highly conserved phosphotransferase that is present in most living cells. Thymidine Kinase catalyzes the phosphorylation reaction: deoxythymidine + ATP = deoxythymidine 5'-phosphate + ADP; it is thus involved in the reaction chain to introduce deoxythymidine into the DNA. Thymidine kinase is required for the action of many antiviral drugs, such as azidothymidine (AZT), and is also used to select hybridoma cell lines in the production of monoclonal antibodies. Thymidine Kinase has many clinical applications as it is only present in anticipation of cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TK1
Applications Western Blot
Classification Monoclonal
Clone C301
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene TK1
Gene Accession No. P04183
Gene Alias D530002A18Rik; EC 2.7.1.21; KITH; thymidine kinase 1; thymidine kinase 1 soluble isoform; thymidine kinase 1, soluble; thymidine kinase, cytosolic; thymidine kinase-1; Tk; Tk1; Tk-1; Tk1a; Tk1b; TK2
Gene Symbols TK1
Host Species Mouse
Immunogen Recombinant Human TK1 protein, Ser2-Asn234 (Accession #P04183).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7083
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.