Learn More
TK1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67787-1-IG-150UL
This item is not returnable.
View return policy
Description
The TK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TK1. This antibody recognizes human antigen. The TK1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| TK1 | |
| Monoclonal | |
| 1000 μg/mL | |
| TK2 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 25 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1B1C2 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 7083 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |