missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TL 13-27

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Negative control for TL 12-186 (Cat. No. 6524)
TL 13-27 is a negative control for TL 12-186 (Cat. No. 6524). Demonstrates no kinase degradation in vitro.
Specifications
Specifications
| Chemical Name or Material | N-(2-(2-(2-(4-(4-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)phenyl)piperazin-1-yl)ethoxy)ethoxy)ethyl)-2-((1,3-dioxo-2-(2-oxopiperidin-3-yl)isoindolin-4-yl)amino)acetamide |
| CAS | 2250025-90-0 |
| Quantity | 5 mg |
| Molecular Formula | C44H53ClN10O8S |
| Formula Weight | 917.48 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction