missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TL1A/TNFSF15 Antibody (2116A) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB74422JF669
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal antibody specifically detects TL1A/TNFSF15 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Neutralization,CyTOF
Specifications
| TL1A/TNFSF15 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
| E. coli-derived recombinant human TL1A/TNFSF15, Leu72-Leu251, Accession # O95150 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot, Neutralization, CyTOF | |
| 2116A | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
| 9966 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction