missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TL1A/TNFSF15 Antibody (2116C), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB7442DL594
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal antibody specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot
Specifications
| TL1A/TNFSF15 | |
| Monoclonal | |
| DyLight 594 | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
| Partial recombinant human TL1A protein (amino acids 72-251) [UniProt O95150] | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
| 9966 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 2116C | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction