missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLCD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06501MFV450
This item is not returnable.
View return policy
Description
TLCD1 Polyclonal antibody specifically detects TLCD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TLCD1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| TLC domain containing 1, TLC domain-containing protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human TLCD1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 116238 | |
| Store at 4C in the dark. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?