missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLDC1 Antibody (OTI5D9), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72091AF750
This item is not returnable.
View return policy
Description
TLDC1 Monoclonal specifically detects TLDC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TLDC1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MEAK7 | |
| Full length recombinant protein of human KIAA1609 (NP_065998) produced in HEK293T cells. | |
| 0.1 mL | |
| 57707.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5D9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| hypothetical protein LOC57707, KIAA1609, MGC25024 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction