Learn More
TLE1 Monoclonal Antibody (OTI1A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800302
Encompass_Preferred
 View more versions of this product
Product Code. 17538032
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with domit-negative AES. Unusual function as coactivator for ESRRG.
Specifications
Specifications
TLE1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q04724
TLE1
Human recombit protein fragment corresponding to amino acids 180-460 of human TLE1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1A11
Unconjugated
TLE1
ESG, ESG1, GRG1
Mouse
Affinity Chromatography
RUO
7088
-20° C, Avoid Freeze/Thaw Cycles