Learn More
Invitrogen™ TLE1 Monoclonal Antibody (OTI1H2)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526424
Product Code. 15916024
  • € 588.00 / 100µL
Description
TLE1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES. Unusual function as coactivator for ESRRG.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLE1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q04724
TLE1
Human recombinant protein fragment corresponding to amino acids 180-460 of TLE1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI1H2
Unconjugated
TLE1
C230057C06Rik; E(Sp1) homolog; enhancer of split groucho; enhancer of split groucho-like protein 1; esg; ESG1; Estm14; grg1; Grg-1; Groucho-related protein 1; RP11-203D19.1; TLE 1; TLE1; tle1.S; tle3b; Tle4l; transducin like enhancer of split 1; transducin like enhancer of split 1 S homeolog; transducin-like enhancer of split 1 (E(sp1) homolog); transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 1, homolog of Drosophila E(spl); transducin-like enhancer of split 3 (E(sp1) homolog) b; transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer protein 1; XELAEV_18001139mg; xgrg1
Mouse
Affinity chromatography
RUO
7088
-20°C, Avoid Freeze/Thaw Cycles