missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1 Monoclonal Antibody (OTI2D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800318
This item is not returnable.
View return policy
Description
Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with domit-negative AES. Unusual function as coactivator for ESRRG.Specifications
| TLE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q04724 | |
| TLE1 | |
| Human recombit protein fragment corresponding to amino acids 180-460 of human TLE1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D8 | |
| Unconjugated | |
| TLE1 | |
| ESG, ESG1, GRG1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7088 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction