Learn More
TLE1 Monoclonal Antibody (OTI2D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800318S
Encompass_Preferred
 View more versions of this product
Product Code. 17568052
  • € 242.00 / 30µL
Description
TLE1 Monoclonal antibody specifically detects TLE1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
TLE1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13546
Mouse
Affinity chromatography
RUO
8737
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI2D8
Unconjugated
RIPK1
C230057C06Rik; E(Sp1) homolog; enhancer of split groucho; enhancer of split groucho-like protein 1; esg; ESG1; Estm14; grg1; Grg-1; Groucho-related protein 1; RP11-203D19.1; TLE 1; TLE1; tle1.S; tle3b; Tle4l; transducin like enhancer of split 1; transducin like enhancer of split 1 S homeolog; transducin-like enhancer of split 1 (E(sp1) homolog); transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 1, homolog of Drosophila E(spl); transducin-like enhancer of split 3 (E(sp1) homolog) b; transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer protein 1; XELAEV_18001139mg; xgrg1
Human recombinant protein fragment corresponding to amino acids 133-422 of human RIPK1 produced in E.coli.
30 μL