missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE2 Antibody (OTI5A6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74531F
This item is not returnable.
View return policy
Description
TLE2 Monoclonal specifically detects TLE2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| TLE2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TLE2 | |
| Full length recombinant protein of human TLE2 (NP_003251) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI5A6 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| enhancer of split groucho 2, ESG, ESG2Enhancer of split groucho-like protein 2, FLJ41188, GRG2, transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 2, homolog of Drosophila E(sp1), transducin-like enhancer protein 2 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 7089.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction