missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLE6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109241
This item is not returnable.
View return policy
Description
Positive Control: Mouse ovary tissue lysate, HepG2, A431, A549, SK-Br-3, mouse small intestine tissue, Daudi.
As a member of the subcortical maternal complex (SCMC), plays an essential role for zygotes to progress beyond the first embryonic cell divisions.
Specifications
| TLE6 | |
| Polyclonal | |
| Unconjugated | |
| TLE6 | |
| 1810057E06Rik; Grg6; Groucho-related protein 6; PREMBL; TLE6; transducin like enhancer of split 6; transducin-like enhancer of split 6 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 6, homolog of Drosophila E(spl); transducin-like enhancer protein 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114606, 79816 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9H808, Q9WVB3 | |
| TLE6 | |
| Synthetic peptide within mouse Tle6 aa 157-206. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction