Learn More
TLK2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99818PCP
Product Code. 18650759
  • € 407.00 / 0.10mL
Description
TLK2 Polyclonal antibody specifically detects TLK2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA
Produced in rabbits immunized with purified, recombinant Human TLK2 (Uniprot#: Q86UE8-1; Leu397-Asn772)
RUO
Human
Polyclonal
0.1 mL
Cell Cycle and Replication