missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLK2 Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805507
This item is not returnable.
View return policy
Description
TLK2, a member of the Ser/Thr protein kinase family, is rapidly and transiently inhibited by phosphorylation following the generation of DNA double-stranded breaks during S-phase. This is cell cycle checkpoint and ATM-pathway dependent and appears to regulate processes involved in chromatin assembly. The TLK2 enzyme is cell-cycle regulated, with maximal activity in S-phase. It is inactivated by phosphorylation at Ser-750, potentially by CHK1. TLK2 heterodimerizes with TLK1. This nuclear protein is widely expressed, with presence in fetal placenta, liver, kidney, pancreas, heart and skeletal muscle tissues, and in several adult cell lines.Specifications
| TLK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86UE8 | |
| TLK2 | |
| Human recombit protein fragment corresponding to amino acids 1-296 of human TLK2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI3E10 | |
| Unconjugated | |
| TLK2 | |
| HsHPK, PKU-ALPHA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11011 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction