missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TLL1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566344

Product Code. 15929054

  • € 580.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GLDYDYTFDGN EEDKTETIDY KDPCKAA Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

This gene encodes an astacin-like zinc-dependent metalloprotease and is a subfamily member of the metzincin family. A similar protein in mice is required during heart development and specifically processes procollagen C-propeptides and chordin at similar cleavage sites.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TLL1
Polyclonal
Unconjugated
TLL1
ASD6; TLL; TLL1; tolloid like 1; tolloid-like 1; tolloid-like protein 1
Rabbit
Antigen affinity chromatography
RUO
7092
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O43897
TLL1
Recombinant Human TLL1. Recombinant protein control fragment (Product #RP-107028).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.