Learn More
Novus Biologicals™ TLR (Intracellular) Screening Antibody Pack
For Research applications
Brand: Novus Biologicals™ NBP2-25085
Includes:
- 1.NBP2-24729: TLR9 Antibody (26C593.2)
- 2.NBP2-24875: TLR3 Antibody (40C1285.6)
- 3.NBP2-24906: TLR7 Antibody
- 4.NBP2-24917: TLR8 Antibody (44C143)
Description
Specifications
| TLR (Intracellular) Screening | |
| Unconjugated | |
| TLR3: 25 ug: WB, Flow, IP, IHC: H: Purified TLR7: 25 ug: WB, Flow, IHC, IP, IF/ICC: H, M: Purified TLR8: 25 ug: WB, Flow, IHC: H, M: Purified TLR9: 25 ug: WB, Flow, IHC, IF/ICC, CM: H, M, R, RM: Purified WB:Western Blot IHC:Immunohistochemistry IP:Immunoprecipitation CM:Confocal Microscopy FA-Neut Functional Assay Neutralization IF/ICC:Immunofluorescence/Immunocytochemistry D:Dog H:Human M:Mouse Mk:Monkey R:Rat RM:Rhesus Monkey | |
| 1.NBP2-24729: TLR9 Antibody (26C593.2) 2.NBP2-24875: TLR3 Antibody (40C1285.6) 3.NBP2-24906: TLR7 Antibody 4.NBP2-24917: TLR8 Antibody (44C143) |
|
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Flow (Intracellular) | |
| 4 Vials | |
| Human, Mouse |
Product Content Correction
Product Title
Spot an opportunity for improvement?Partager une correction de contenu