Learn More
Novus Biologicals™ TLR Screening Antibody Pack
For Research applications
Brand:  Novus Biologicals™ NBP2-25083
Product Code. 18766673
  • ONLINE EXCLUSIVE PRICE
    € 1290.00
    € 1220.10 / Pack of 11
    Save € 69.90
    5% Off
Includes:
  • 1.NB100-56563: TLR1 Antibody
  • 2.NB100-56720: TLR2 Antibody
  • 3.NB100-56722: TLR2 Antibody (TL2.1)
  • 4.NB100-56723: TLR4 Antibody (HTA125)
  • 5.NB100-56728: TLR2 Antibody (1030A5.138)
  • 6.NBP2-24729: TLR9 Antibody (26C593.2)
  • 7.NBP2-24827: TLR5 Antibody
  • 8.NBP2-24875: TLR3 Antibody (40C1285.6)
  • 9.NBP2-24906: TLR7 Antibody
  • 10.NBP2-24917: TLR8 Antibody (44C143)