missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR11 Antibody (786404), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7640N
This item is not returnable.
View return policy
Description
TLR11 Monoclonal antibody specifically detects TLR11 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TLR11 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 786404 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| hgnc31753, Toll like receptor 11 | |
| E. coli-derived recombinant mouse TLR11, Thr209-Gln325, Accession # Q6R5P0 | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 442887 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction