missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (11G5), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-27166AF350
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TLR2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TLR2 | |
| A portion of amino acids 400-450 of murine TLR2 was used as the immunogen for this antibody. | |
| Primary | |
| Mouse | |
| IgG2a κ |
| Western Blot, Flow Cytometry | |
| 11G5 | |
| Western Blot, Flow Cytometry | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Rat | |
| 0.1 mL | |
| 7097 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction