missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (11G5), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-27165AF488
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TLR2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 85% homologous in Vole, 80% in Rat, Chinese Hamster and Rabbit, 70% in Orangutan, Rhesus Monkey and Horse, and 65% homologous in Human. | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry | |
| 11G5 | |
| Western Blot, Flow Cytometry | |
| TLR2 | |
| A portion of amino acids 400-450 of murine TLR2 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7097 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction