missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (11G5) - Azide Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-27166
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| TLR2 | |
| Monoclonal | |
| 0.5mg/mL | |
| Western Blot 4-6ug/ml∼, Flow Cytometry 1ul/1 million cells, CyTOF-ready | |
| TLR2 | |
| A portion of amino acids 400-450 of murine TLR2 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in Mouse, 85% in Vole, 80% in Rat, Chinese Hamster and Rabbit, 70% in Orangutan, Rhesus Monkey and Horse, and 65% homologous in Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, CyTOF | |
| 11G5 | |
| Unconjugated | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 7097 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction