missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (203325), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Bio-Techne FAB1530Z
This item is not returnable.
View return policy
Description
TLR2 Monoclonal antibody specifically detects TLR2 in Mouse samples. It is validated for Flow Cytometry
Specifications
| TLR2 | |
| Monoclonal | |
| DyLight 755 | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TLR2, Gln25-Leu590, Accession # Q9QUN7 | |
| 0.5 mL | |
| Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 7097 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 203325 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction