missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (67N8F8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25259APC
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Flow Cytometry.
Specifications
| TLR2 | |
| Monoclonal | |
| APC | |
| NP_003255.2 | |
| TLR2 | |
| Amino acids 227-371 of human TLR2 were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7097 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 67N8F8 | |
| Flow Cytometry | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction