missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody (27N3D4), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-27405DL594
This item is not returnable.
View return policy
Description
TLR3 Monoclonal antibody specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| TLR3 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 27N3D4 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| 7098 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction