missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody (40C1285.6), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24875AF488
This item is not returnable.
View return policy
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| TLR3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| O15455 | |
| TLR3 | |
| A synthetic peptide corresponding to amino acids 55-85 of human TLR3 was used as immunogen. | |
| 0.1 mL | |
| Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7098 | |
| Human, Mouse, Canine | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 40C1285.6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Dog, Human and Mouse. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction