missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody (40C1285.6), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24875JF549
This item is not returnable.
View return policy
Description
TLR3 Monoclonal antibody specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow (Intracellular), Bioactivity, Neutralization
Specifications
| TLR3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| A synthetic peptide corresponding to amino acids 55-85 of human TLR3 was used as immunogen. | |
| 0.1 mL | |
| Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| 7098 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 40C1285.6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Canine | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu