missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Antibody (610015), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6248MFV450
This item is not returnable.
View return policy
Description
TLR4 Monoclonal antibody specifically detects TLR4 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TLR4 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Autophagy, Cytokine Research, Immunology, Inflammation, Innate Immunity, Neurodegeneration, Signal Transduction, Virology Bacteria and Parasites | |
| 7099 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 610015 | |
| Flow Cytometry, CyTOF-ready | |
| ARMD10, CD_antigen: CD284, CD284, CD284 antigen, EC 3.2.2.6, EC:3.2.2.6, homolog of Drosophila toll, hToll, TLR4, TLR-4, TOLL, toll like receptor 4 protein, toll-like receptor 4 | |
| Mouse myeloma cell line NS0-derived recombinant human TLR4, Glu24-Lys631, Accession # O00206 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction