missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Antibody (76B357.1), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27149FR
This item is not returnable.
View return policy
Description
TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface), Knockout Validated, Knockdown Validated.
Specifications
| TLR4 | |
| Monoclonal | |
| DyLight 680 | |
| O00206 | |
| TLR4 | |
| This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564). | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| 7099 | |
| Human, Mouse, Rat, Pig, Bovine, Mammalia | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| 76B357.1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface), Knockout Validated, KnockDown Validated | |
| ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used has 100% identity in human, chimp, baboon, Chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. Human, Mouse, Rat. Ground squirrel reactivity reported by a customer review. Predicted cross-reactivity based on sequence identity: Chimp (100%), Baboon (100%), Chinese Hamster (100%), Bovine (93%), Pig (93%). Mammal reactivity reported in scientific literature (PMID: 25130694). | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction