missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Antibody (TLR4/3895R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08732AF405
This item is not returnable.
View return policy
Description
TLR4 Monoclonal antibody specifically detects TLR4 in Human, Rat, Porcine, Canine, Guinea Pig, Monkey samples. It is validated for ELISA
Specifications
| TLR4 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| ARMD10, CD_antigen: CD284, CD284, CD284 antigen, EC 3.2.2.6, EC:3.2.2.6, homolog of Drosophila toll, hToll, TLR4, TLR-4, TOLL, toll like receptor 4 protein, toll-like receptor 4 | |
| Recombinant full-length human TLR4 protein. (Uniprot: O00206) | |
| 100 μg | |
| Adaptive Immunity, Autophagy, Cytokine Research, Immunology, Inflammation, Innate Immunity, Neurodegeneration, Signal Transduction, Virology Bacteria and Parasites | |
| 7099 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| TLR4/3895R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Canine, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction