missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen
This item is not returnable.
View return policy
Description
TLR4 Polyclonal antibody specifically detects TLR4 in Human samples. It is validated for ELISA, Western Blot.
Specifications
| TLR4 | |
| Polyclonal | |
| Unconjugated | |
| TLR4 | |
| ARMD10; CD284; EGK_07381; homolog of Drosophila toll; hToll; lipopolysaccharide response; Lps; Ly87; Ran/M1; Rasl2-8; Tlr4; TLR-4; TOLL; toll like receptor 4; toll/interleukin-l receptor (TIR) domain; toll4; toll-like receptor 4; toll-like receptor 4 immunity-related protein; Toll-like receptor 4-like protein; Toll-like receptor4 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7099 | |
| -20°C | |
| Liquid |
| ELISA, Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| O00206 | |
| TLR4 | |
| Synthetic peptide derived from the carboxyl termini of various human TLR4 (Toll-like receptor 4) isoforms A-D | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction