missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR5 Antibody (85B152.5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97728AF700
This item is not returnable.
View return policy
Description
TLR5 Monoclonal specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).
Specifications
| TLR5 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| O60602 | |
| TLR5 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5. It will cross-react with mouse TLR5 (NP_003259). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Canine | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 85B152.5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular) | |
| MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5 | |
| Mouse | |
| 100 kDa | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
| 7100 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction