missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLR5 Monoclonal Antibody (8-1#)

Mouse Monoclonal Antibody
€ 180.00 - € 478.00
Specifications
| Antigen | TLR5 |
|---|---|
| Clone | 8-1# |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17887802
![]() |
Invitrogen™
MA541682 |
200 μL |
€ 478.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17674156
|
Invitrogen™
MA543703 |
20 μL |
€ 180.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR5 Monoclonal Antibody for Western Blot, IHC (P)
TLR5 is a Type I membrane protein belonging to the Toll-like receptor family, participates in the innate immune response to microbial agents. TLR5 also plays a role in mediating detection of bacterial flagellins. TLR5 acts via MyD88 and TRAF6, leading to NF-kappa-B activation, cytokine secretion and the inflammatory response. TLR5 protein binds to TIRAP and MyD88 via their respective TIR domains TLR5 is highly expressed in liver, and is detected in lung and at very low levels in most other tissues. The TLR5 gene lies in a locus that is associated with susceptibility to Salmonella. Inbred strains of mice can be classified into 3 categories according to their resistance to infection with S.typhimurium: susceptible (BALB/c, C57BL/6, C3H/He), intermediate (DBA/2, C75L) and resistant (A, CBA). The strain MOLF/Ei is highly susceptible to the infection, has a unique TLR5 haplotype and a lower expression of TRL5. A common TLR5 stop codon polymorphism that disrupts TLR5 signaling is associated with susceptibility to Legionnaires' disease and demonstrates the importance of TLR5 in the innate immune response.Specifications
| TLR5 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O60602 | |
| 289337, 7100 | |
| Recombinant protein Toll Like Receptor 5. The antigen corresponds to amino acid range 693-858 of the target protein. | |
| Antibody |
| 8-1# | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human, Rat | |
| TLR5 | |
| FLJ10052; MELIOS; MGC126430; MGC126431; RP11-239E10.1; SLE1; SLEB1; systemic lupus erythematosus susceptibility 1; TIL3; Tlr5; toll like receptor 5; Toll/interleukin-1 receptor-like protein 3; toll-like receptor 5 | |
| TLR5 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title