missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR6 Antibody (418601) [Janelia Fluor™ 646], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1533J
This item is not returnable.
View return policy
Description
TLR6 Monoclonal antibody specifically detects TLR6 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TLR6 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 418601 | |
| Flow Cytometry, CyTOF-ready | |
| CD286, CD286 antigen, toll-like receptor 6 | |
| HEK293 human embryonic kidney cell line transfected with mouse TLR6, Phe39-Thr806, Accession # BAA78632 | |
| 0.1 mL | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer | |
| 10333 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction