missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR6 Antibody (86B1153.2), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37704JF549
This item is not returnable.
View return policy
Description
TLR6 Monoclonal specifically detects TLR6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TLR6 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TLR6 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6. | |
| 0.1 mL | |
| Primary | |
| TLR6 (86B1153.2) | |
| Store at 4°C in the dark. |
| Immunohistochemistry (Paraffin) | |
| 86B1153.2 | |
| Immunohistochemistry-Paraffin | |
| CD286, CD286 antigen, toll-like receptor 6 | |
| Mouse | |
| Protein G purified | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
| 10333.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction