Learn More
TLR6 Antibody (TLR6.127), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51493
This item is not returnable.
View return policy
Description
TLR6 Monoclonal specifically detects TLR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| TLR6 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| TLR6 | |
| TLR6.127 reacts with the human Toll-like receptor 6 | |
| 0.05 mg | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
| 10333 | |
| Store at 4°C. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| TLR6.127 | |
| Unconjugated | |
| CD286, CD286 antigen, toll-like receptor 6 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |