missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody (4F4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04349B
This item is not returnable.
View return policy
Description
TLR7 Monoclonal specifically detects TLR7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| TLR7 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TLR7 | |
| Recombinant human TLR7 (451-500aa) purified from E. coli | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| 51284 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4F4 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| toll-like receptor 7 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human TLR7 mAb, clone 4F4, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TLR7 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction