missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR7 Antibody (4G6), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27332APCCY7
This item is not returnable.
View return policy
Description
TLR7 Monoclonal specifically detects TLR7 in Human, Mouse samples. It is validated for Flow Cytometry, Knockdown Validated.
Specifications
| TLR7 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TLR7 | |
| A partial human TLR7 recombinant protein (amino acids 562-839) was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 κ |
| Flow Cytometry, KnockDown | |
| 4G6 | |
| Flow Cytometry, KnockDown Validated | |
| toll-like receptor 7 | |
| Mouse | |
| Protein G purified | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| 51284.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction